2-(2-Trifluoromethylbenzoimidazol-1-ylpropionic acid hydrochloride
Supplier: Matrix Scientific
034315-500MG
101837-410EA
153.85
USD
101837-410
2-(2-Trifluoromethylbenzoimidazol-1-ylpropionic acid hydrochloride
2-(2-Trifluoromethylbenzoimidazol-1-ylpropionic acid hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₀ClF₃N₂O₂ |
MDL Number:
MFCD11506596 CAS Number: 1219422-80-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...