Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(2-Amino-thiazol-4-yl)-acetamide

Supplier: Matrix Scientific


034305-500MG
101837-390EA 261.54 USD InStock
101837-390
2-(2-Amino-thiazol-4-yl)-acetamide
2-(2-Aminothiazol-4-yl)acetamide
CAS: 220041-33-8
MDL Number: MFCD07689461
Molecular Formula: C5H7N3OS
Molecular Weight: 157,2
Hazard: Irritant


Formula: C₅H₇N₃OS
MW: 157,2 g/mol
MDL Number: MFCD07689461
CAS Number: 220041-33-8
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty