5-(Piperidine-1-sulfonyl)-2,3-dihydro-1H-indole

Supplier: Matrix Scientific

034304-500MG
101837-388EA 246.15 USD
101837-388
5-(Piperidine-1-sulfonyl)-2,3-dihydro-1H-indole
5-(Piperidine-1-sulfonyl)-2,3-dihydro-1H-indole

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂S
MDL Number: MFCD07687988
CAS Number: 874593-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR