2-Methylimidazo[1,2-a]pyridine-3-carboxylic acid

Supplier: Matrix Scientific

034302-500MG
101837-384EA 230 USD
101837-384
2-Methylimidazo[1,2-a]pyridine-3-carboxylic acid
2-Methylimidazo[1,2-a]pyridine-3-carboxylic acid

Hazard: Irritant

Formula: C₉H₈N₂O₂
MW: 176.1700 g/mol
Boiling Pt: 170 °C
Melting Pt: 140 °C
Density: 1.359
MDL Number: MFCD00244615
CAS Number: 21801-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR