(4-sulfamoylphenoxy)acetic acid

Supplier: Matrix Scientific

034293-500MG
101837-366EA 307.69 USD
101837-366
(4-sulfamoylphenoxy)acetic acid
(4-sulfamoylphenoxy)acetic acid

Hazard: Irritant

Formula: C₈H₉NO₅S
MDL Number: MFCD07400614
CAS Number: 7383-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR