2-Amino-6-ethyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylic acid amide

Supplier: Matrix Scientific

034285-500MG
101837-348EA 384.62 USD
101837-348
2-Amino-6-ethyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylic acid amide
2-Amino-6-ethyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylic acid amide

Hazard: Irritant

Formula: C₁₀H₁₅N₃OS
MDL Number: MFCD01827706
CAS Number: 355001-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR