3-(Cyclopentyloxy)aniline

Supplier: Matrix Scientific

034277-500MG
101837-332EA 204.62 USD
101837-332
3-(Cyclopentyloxy)aniline
3-(Cyclopentyloxy)aniline

Hazard: Irritant

Formula: C₁₁H₁₅NO
MDL Number: MFCD09036966
CAS Number: 653604-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR