5,6,7,8-Tetrahydro-4H-cyclohepta[d]isoxazole-3-carboxylic acid

Supplier: Matrix Scientific

034274-500MG
101837-326EA 292.31 USD
101837-326
5,6,7,8-Tetrahydro-4H-cyclohepta[d]isoxazole-3-carboxylic acid
5,6,7,8-Tetrahydro-4H-cyclohepta[d]isoxazole-3-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₁NO₃
MDL Number: MFCD04969900
CAS Number: 33230-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR