4-(Pyrrolidine-1-sulfonyl)benzylamine

Supplier: Matrix Scientific

034273-500MG
101837-324EA 384.62 USD
101837-324
4-(Pyrrolidine-1-sulfonyl)benzylamine
4-(Pyrrolidine-1-sulfonyl)benzylamine

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂S
MDL Number: MFCD08337820
CAS Number: 784997-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR