4-p-Tolyl-2H-pyrazol-3-ylamine hydrochloride
Supplier: Matrix Scientific
034251-500MG
101837-280EA
307.69
USD
101837-280
4-p-Tolyl-2H-pyrazol-3-ylamine hydrochloride
4-p-Tolyl-2H-pyrazol-3-ylamine hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₂ClN₃ |
MDL Number:
MFCD11506591 CAS Number: 1239866-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...