[2-(3-Aminomethylphenoxy)ethyl]dimethylamine

Supplier: Matrix Scientific

034232-500MG
101837-242EA 461.54 USD
101837-242
[2-(3-Aminomethylphenoxy)ethyl]dimethylamine
[2-(3-Aminomethylphenoxy)ethyl]dimethylamine

Hazard: Irritant

Formula: C₁₁H₁₈N₂O
MDL Number: MFCD09043583
CAS Number: 140836-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR