3-(4-Fluorobenzyloxy)phenylamine
Supplier: Matrix Scientific
034212-500MG
101837-204EA
246.15
USD
101837-204
3-(4-Fluorobenzyloxy)phenylamine
3-(4-Fluorobenzyloxy)phenylamine
Hazard: Irritant
Formula:
C₁₃H₁₂FNO |
MDL Number:
MFCD07801209 CAS Number: 79808-10-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...