2,2-Dimethyl-3-(4-methylpiperazin-1-yl)propylamine ≥95%

Supplier: Matrix Scientific

034178-500MG
101837-136EA 461.54 USD
101837-136
2,2-Dimethyl-3-(4-methylpiperazin-1-yl)propylamine ≥95%
2,2-Dimethyl-3-(4-methylpiperazin-1-yl)propylamine

Hazard: Irritant

Formula: C₁₀H₂₃N₃
MW: 185.31 g/mol
MDL Number: MFCD06740639
CAS Number: 879059-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR