5-(2-Chlorobenzyl)thiazol-2-ylamine

Supplier: Matrix Scientific

034166-500MG
101837-112EA 461.54 USD
101837-112
5-(2-Chlorobenzyl)thiazol-2-ylamine
5-(2-Chlorobenzyl)thiazol-2-ylamine

Hazard: Irritant

Formula: C₁₀H₉ClN₂S
MW: 224.71 g/mol
MDL Number: MFCD00667685
CAS Number: 207463-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR