3-Fluoro-4-pyrrolidin-1-ylphenylamine dihydrochloride

Supplier: Matrix Scientific

034165-500MG
101837-110EA 307.69 USD
101837-110
3-Fluoro-4-pyrrolidin-1-ylphenylamine dihydrochloride
3-Fluoro-4-pyrrolidin-1-ylphenylamine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₅Cl₂FN₂
MDL Number: MFCD08087469
CAS Number: 93246-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR