2,5-Dimethyl-1-pyridin-2-ylmethyl-1H-pyrrole-3-carbaldehyde

Supplier: Matrix Scientific

034164-500MG
101837-108EA 307.69 USD
101837-108
2,5-Dimethyl-1-pyridin-2-ylmethyl-1H-pyrrole-3-carbaldehyde
2,5-Dimethyl-1-pyridin-2-ylmethyl-1H-pyrrole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₄N₂O
MDL Number: MFCD07791605
CAS Number: 879053-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR