4-(Morpholine-4-sulfonyl)benzylamine

Supplier: Matrix Scientific

034161-500MG
101837-102EA 461.54 USD
101837-102
4-(Morpholine-4-sulfonyl)benzylamine
4-(Morpholine-4-sulfonyl)benzylamine

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₃S
MDL Number: MFCD06655508
CAS Number: 852399-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR