3-Ethoxy-4-pyrrolidin-1-ylbenzaldehyde

Supplier: Matrix Scientific

034157-500MG
101837-094EA 461.54 USD
101837-094
3-Ethoxy-4-pyrrolidin-1-ylbenzaldehyde
3-Ethoxy-4-pyrrolidin-1-ylbenzaldehyde

Hazard: Irritant

Formula: C₁₃H₁₇NO₂
MDL Number: MFCD07403410
CAS Number: 879047-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR