2-Fluoro-4-piperidin-1-ylbenzaldehyde

Supplier: Matrix Scientific

034154-500MG
101837-088EA 307.69 USD
101837-088
2-Fluoro-4-piperidin-1-ylbenzaldehyde
2-Fluoro-4-piperidin-1-ylbenzaldehyde

Hazard: Irritant

Formula: C₁₂H₁₄FNO
MDL Number: MFCD06740033
CAS Number: 245725-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR