5-Isopropyl-4-methoxy-2-methyl-benzylamine

Supplier: Matrix Scientific

034152-500MG
101837-084EA 307.69 USD
101837-084
5-Isopropyl-4-methoxy-2-methyl-benzylamine
5-Isopropyl-4-methoxy-2-methyl-benzylamine

Hazard: Irritant

Formula: C₁₂H₁₉NO
MDL Number: MFCD07403241
CAS Number: 879063-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR