2-Bromo-1-(4-methoxy-2,5-dimethylphenyl)ethanone

Supplier: Matrix Scientific

034147-500MG
101837-074EA 461.54 USD
101837-074
2-Bromo-1-(4-methoxy-2,5-dimethylphenyl)ethanone
2-Bromo-1-(4-methoxy-2,5-dimethylphenyl)ethanone

Hazard: Irritant

Formula: C₁₁H₁₃BrO₂
MW: 257.13 g/mol
MDL Number: MFCD07403173
CAS Number: 879053-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR