5-Chloro-2-ethoxy-nicotinic acid

Supplier: Matrix Scientific

Synonyms: 5-Chloro-2-ethoxynicotinic acid

034146-500MG
101837-072EA 307.69 USD
101837-072
5-Chloro-2-ethoxy-nicotinic acid
5-Chloro-2-ethoxy-nicotinic acid

Hazard: Irritant

Formula: C₈H₈ClNO₃
MW: 201.61 g/mol
MDL Number: MFCD07403147
CAS Number: 68359-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR