1-(2-Methoxyethyl)-1H-benzoimidazole-5-carboxylic acid

Supplier: Matrix Scientific

034111-500MG
101837-004EA 307.69 USD
101837-004
1-(2-Methoxyethyl)-1H-benzoimidazole-5-carboxylic acid
1-(2-Methoxyethyl)-1H-benzoimidazole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃
MDL Number: MFCD07790838
CAS Number: 284672-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR