5-(4-Chlorophenyl)-2-methyl-2H-pyrazole-3-carboxylic acid
Supplier: Matrix Scientific
Synonyms:
3-(4-Chlorophenyl)-1-methyl-1H-pyrazole-5-carboxylic acid
034105-500MG
101836-992EA
461.54
USD
101836-992
5-(4-Chlorophenyl)-2-methyl-2H-pyrazole-3-carboxylic acid
5-(4-Chlorophenyl)-2-methyl-2H-pyrazole-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₉ClN₂O₂ MW: 236.66 g/mol |
MDL Number:
MFCD08446011 CAS Number: 1015868-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...