2-Ethyl-5-phenyl-2H-pyrazole-3-carboxylic acid

Supplier: Matrix Scientific

034104-500MG
101836-990EA 461.54 USD
101836-990
2-Ethyl-5-phenyl-2H-pyrazole-3-carboxylic acid
2-Ethyl-5-phenyl-2H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂
MDL Number: MFCD08445971
CAS Number: 10199-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR