5-(2,4-Dimethoxyphenyl)-4-methyl-2H-pyrazole-3-carboxylic acid

Supplier: Matrix Scientific

034089-500MG
101836-960EA 246.15 USD
101836-960
5-(2,4-Dimethoxyphenyl)-4-methyl-2H-pyrazole-3-carboxylic acid
5-(2,4-Dimethoxyphenyl)-4-methyl-2H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₄
MW: 262.27 g/mol
MDL Number: MFCD07400763
CAS Number: 890625-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR