4-(4-Fluoro-3-methylphenyl)butyric acid

034076-500MG
101836-934EA 261.05 USD
101836-934
4-(4-Fluoro-3-methylphenyl)butyric acid
4-(4-Fluoro-3-methylphenyl)butyric acid

Hazard: Irritant

Formula: C₁₁H₁₃FO₂
MW: 196.22 g/mol
MDL Number: MFCD07790543
CAS Number: 331-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR