4-[2-(3,4-Dimethoxyphenyl)ethyl]piperidine

Supplier: Matrix Scientific

034051-500MG
101836-884EA 246.15 USD
101836-884
4-[2-(3,4-Dimethoxyphenyl)ethyl]piperidine
4-[2-(3,4-Dimethoxyphenyl)ethyl]piperidine

Hazard: Irritant

Formula: C₁₅H₂₃NO₂
MDL Number: MFCD05022479
CAS Number: 893755-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR