1-Methyl-1H-indol-5-ylamine

Supplier: Matrix Scientific

Synonyms: 1-Methyl-1H-indol-5-amine5-Amino-1-methyl-1H-indole

034047-500MG
101836-876EA 152.31 USD
101836-876
1-Methyl-1H-indol-5-ylamine
1-Methyl-1H-indol-5-ylamine

Hazard: Irritant

Formula: C₉H₁₀N₂
MW: 146.19 g/mol
MDL Number: MFCD03839873
CAS Number: 102308-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR