2,4-Dimethyl-6-piperazin-1-yl-pyrimidine

Supplier: Matrix Scientific

034038-500MG
101836-858EA 216.92 USD
101836-858
2,4-Dimethyl-6-piperazin-1-yl-pyrimidine
2,4-Dimethyl-6-piperazin-1-yl-pyrimidine

Hazard: Irritant

Formula: C₁₀H₁₆N₄
MDL Number: MFCD07365286
CAS Number: 887686-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR