2-Methylbenzothiazole-6-sulfonyl chloride

Supplier: Matrix Scientific

034035-500MG
101836-852EA 384.62 USD
101836-852
2-Methylbenzothiazole-6-sulfonyl chloride
2-Methylbenzothiazole-6-sulfonyl chloride

Hazard: Irritant

Formula: C₈H₆ClNO₂S₂
MW: 247.72 g/mol
MDL Number: MFCD03990571
CAS Number: 21431-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR