Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Pyrrolidin-1-yl-benzylamine

Supplier: Matrix Scientific


034017-500MG
101836-816EA 261.54 USD InStock
101836-816
2-Pyrrolidin-1-yl-benzylamine
2-Pyrrolidin-1-ylbenzylamine
CAS: 72752-53-5
MDL Number: MFCD04971089
Molecular Formula: C11H16N2
Molecular Weight: 176,26
Hazard: Irritant


Formula: C₁₁H₁₆N₂
MW: 176.26 g/mol
MDL Number: MFCD04971089
CAS Number: 72752-53-5
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty