2-[4-(2-Aminophenyl)piperazin-1-yl]ethanol

Supplier: Matrix Scientific

034012-500MG
101836-806EA 461.54 USD
101836-806
2-[4-(2-Aminophenyl)piperazin-1-yl]ethanol
2-[4-(2-Aminophenyl)piperazin-1-yl]ethanol

Hazard: Irritant

Formula: C₁₂H₁₉N₃O
MDL Number: MFCD04971044
CAS Number: 869946-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR