5-Fluoro-2-morpholin-4-ylphenylamine

Supplier: Matrix Scientific

034008-500MG
101836-798EA 461.54 USD
101836-798
5-Fluoro-2-morpholin-4-ylphenylamine
5-Fluoro-2-morpholin-4-ylphenylamine

Hazard: Irritant

Formula: C₁₀H₁₃FN₂O
MW: 196.23 g/mol
MDL Number: MFCD04971024
CAS Number: 144187-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR