4-(1,5-Dimethyl-1H-pyrazol-4-yl)thiazol-2-ylamine
Supplier: Matrix Scientific
033993-500MG
101836-768EA
461.54
USD
101836-768
4-(1,5-Dimethyl-1H-pyrazol-4-yl)thiazol-2-ylamine
4-(1,5-Dimethyl-1H-pyrazol-4-yl)thiazol-2-ylamine
Hazard: Irritant
Formula:
C₈H₁₀N₄S MW: 194.26 g/mol |
MDL Number:
MFCD04969956 CAS Number: 957242-02-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...