4-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)thiazol-2-ylamine

Supplier: Matrix Scientific

033992-500MG
101836-766EA 307.69 USD
101836-766
4-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)thiazol-2-ylamine
4-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)thiazol-2-ylamine

Hazard: Irritant

Formula: C₉H₁₂N₄S
MW: 208.29 g/mol
MDL Number: MFCD04969875
CAS Number: 957301-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR