3-Imidazo[2,1-b]thiazol-6-ylphenylamine

Supplier: Matrix Scientific

033973-500MG
101836-728EA 307.69 USD
101836-728
3-Imidazo[2,1-b]thiazol-6-ylphenylamine
3-Imidazo[2,1-b]thiazol-6-ylphenylamine

Hazard: Irritant

Formula: C₁₁H₉N₃S
MW: 215.28 g/mol
MDL Number: MFCD03617826
CAS Number: 861206-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR