Methyl(2-methyl-2H-pyrazol-3-ylmethyl)amine

Supplier: Matrix Scientific

Synonyms: 1-Methyl-5-[(methylamino)methyl]-1H-pyrazole

033970-500MG
101836-722EA 176.92 USD
101836-722
Methyl(2-methyl-2H-pyrazol-3-ylmethyl)amine
Methyl(2-methyl-2H-pyrazol-3-ylmethyl)amine

Hazard: Irritant

Formula: C₆H₁₁N₃
MW: 125.17 g/mol
MDL Number: MFCD03419812
CAS Number: 930111-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR