(4-Methoxybenzenesulfonyl)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-((4-Methoxyphenyl)sulphonyl)acetic acid2-((4-Methoxyphenyl)sulfonyl)acetic acid

033963-500MG
101836-708EA 384.62 USD
101836-708
(4-Methoxybenzenesulfonyl)acetic acid
(4-Methoxybenzenesulfonyl)acetic acid

Hazard: Irritant

Formula: C₉H₁₀O₅S
MW: 230.24 g/mol
MDL Number: MFCD07364114
CAS Number: 3937-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR