3-(3,5-Dimethyl-1H-pyrazol-4-ylmethyl)-4-methoxybenzaldehyde hydrochloride

Supplier: Matrix Scientific

033955-500MG
101836-692EA 307.69 USD
101836-692
3-(3,5-Dimethyl-1H-pyrazol-4-ylmethyl)-4-methoxybenzaldehyde hydrochloride
3-(3,5-Dimethyl-1H-pyrazol-4-ylmethyl)-4-methoxybenzaldehyde hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₇ClN₂O₂
MDL Number: MFCD11506558
CAS Number: 1033076-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR