4-Allyloxybenzoic acid

Supplier: Matrix Scientific

Synonyms: 4-(Allyloxy)benzoic acid

033951-500MG
101836-684EA 230.77 USD
101836-684
4-Allyloxybenzoic acid
4-Allyloxybenzoic acid

Hazard: Irritant

Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
MDL Number: MFCD00526542
CAS Number: 27914-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR