3-(3-Fluorobenzyloxy)benzoic acid

Supplier: Matrix Scientific

033947-500MG
101836-676EA 307.69 USD
101836-676
3-(3-Fluorobenzyloxy)benzoic acid
3-(3-Fluorobenzyloxy)benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₁FO₃
MDL Number: MFCD07398695
CAS Number: 887599-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR