3',5'-Dimethyl-1'-propyl-1H,1'H-[3,4']bipyrazolyl-5-carboxylic acid

Supplier: Matrix Scientific

033934-500MG
101836-650EA 307.69 USD
101836-650
3',5'-Dimethyl-1'-propyl-1H,1'H-[3,4']bipyrazolyl-5-carboxylic acid
3',5'-Dimethyl-1'-propyl-1H,1'H-[3,4']bipyrazolyl-5-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₆N₄O₂
MDL Number: MFCD07397888
CAS Number: 890621-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR