C-[5-(4-tert-Butylphenyl)[1,2,4]oxadiazol-3-yl]methylamine hydrochloride
Supplier: Matrix Scientific
033924-500MG
101836-632EA
307.69
USD
101836-632
C-[5-(4-tert-Butylphenyl)[1,2,4]oxadiazol-3-yl]methylamine hydrochloride
C-[5-(4-tert-Butylphenyl)[1,2,4]oxadiazol-3-yl]methylamine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₈ClN₃O |
MDL Number:
MFCD11506557 CAS Number: 1185294-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...