5-(3-Methoxyphenyl)-2H-pyrazol-3-ylamine hydrochloride

Supplier: Matrix Scientific

033910-500MG
101836-604EA 307.69 USD
101836-604
5-(3-Methoxyphenyl)-2H-pyrazol-3-ylamine hydrochloride
5-(3-Methoxyphenyl)-2H-pyrazol-3-ylamine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₂ClN₃O
MW: 225.68 g/mol
MDL Number: MFCD05256460
CAS Number: 1025447-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR