3-(Piperidin-1-ylsulfonyl)aniline

Supplier: Matrix Scientific

033884-500MG
101836-552EA 55.38 USD
101836-552
3-(Piperidin-1-ylsulfonyl)aniline
3-(Piperidin-1-ylsulphonyl)aniline

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂S
MDL Number: MFCD02063328
CAS Number: 22184-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR