4-(2-Pyrrolidin-1-ylethoxy)phenylamine

Supplier: Matrix Scientific

Synonyms: 4-(2-(Pyrrolidin-1-yl)ethoxy)aniline

033869-500MG
101836-524EA 338.46 USD
101836-524
4-(2-Pyrrolidin-1-ylethoxy)phenylamine
4-(2-Pyrrolidin-1-ylethoxy)phenylamine

Hazard: Irritant/Stored under Argon

Formula: C₁₂H₁₈N₂O
MW: 206.29 g/mol
MDL Number: MFCD07790198
CAS Number: 50609-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR