4-Methyl-1H-indole-2-carboxylic acid

Supplier: Matrix Scientific

033816-500MG
101836-420EA 160 USD
101836-420
4-Methyl-1H-indole-2-carboxylic acid
4-Methyl-1H-indole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₉NO₂
MW: 175.1800 g/mol
MDL Number: MFCD02664476
CAS Number: 18474-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR