6-Fluoro-1H-indole-2-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 6-Fluoroindole-2-carboxylic acid

033815-500MG
101836-418EA 307.69 USD
101836-418
6-Fluoro-1H-indole-2-carboxylic acid
6-Fluoro-1H-indole-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₆FNO₂
MW: 179.15 g/mol
MDL Number: MFCD01863162
CAS Number: 3093-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR