5-(2,4-Dichlorophenyl)-2H-pyrazol-3-ylamine hydrochloride

Supplier: Matrix Scientific

033812-500MG
101836-414EA 307.69 USD
101836-414
5-(2,4-Dichlorophenyl)-2H-pyrazol-3-ylamine hydrochloride
5-(2,4-Dichlorophenyl)-2H-pyrazol-3-ylamine hydrochloride

Hazard: Irritant

Formula: C₉H₈Cl₃N₃
MW: 264.54 g/mol
MDL Number: MFCD06653176
CAS Number: 1025447-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR